Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 1,2,5-trimethylpyrrole-3-carboxylate, 97%

$21.69 - $209.34

Chemical Identifiers

Chemical Name or Material Methyl 1,2,5-trimethylpyrrole-3-carboxylate
CAS 14186-50-6
Molecular Formula C9H13NO2
Formula Weight 167.21
MDL Number MFCD00203867
Synonym 1,2,5-Trimethyl-1H-pyrrole-3-carboxylic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2548103

  SDS
alfa aesar™
B2548103
1g Each for $21.69

AAB2548114

  SDS
alfa aesar™
B2548114
25g Each for $56.96

AAB2548122

  SDS
alfa aesar™
B2548122
100g Each for $209.34
Description & Specifications

Specifications

Formula Weight 167.21
Assay 97%
CAS 14186-50-6
Synonym 1,2,5-Trimethyl-1H-pyrrole-3-carboxylic acid methyl ester
Health Hazard 1 Warning
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Methyl 1,2,5-trimethylpyrrole-3-carboxylate
Molecular Formula C9H13NO2
MDL Number MFCD00203867
Melting Point 115° to 118°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.