Please login to your online account to display your discounted pricing

Methyl 1,2,3,4-tetrahydroquinoline-2-carboxylate hydrochloride, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material Methyl 1,2,3,4-tetrahydroquinoline-2-carboxylate hydrochloride
CAS 78348-26-2
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H13NO2·ClH
Formula Weight 227.69
MDL Number MFCD02180877
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SPB08364CB

 
fisher scientific company
SPB08364CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 227.69
Quantity 250mg
Assay 97%
CAS Min % 97.0
Molecular Formula C11H13NO2·ClH
CAS 78348-26-2
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Methyl 1,2,3,4-tetrahydroquinoline-2-carboxylate hydrochloride
Packaging Amber Glass Bottle
Assay Percent Range 97%
MDL Number MFCD02180877
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.