Alfa Aesar™ Methoxypolyethylene glycol amine, M.W. 1,000
Manufacturer:  Alfa Aesar™ 4664306
 View more versions of this product
Catalog No. AA4664306
Specifications
Specifications
Methoxypolyethylene glycol amine
80506-64-5
MFCD00131599
MSKSQCLPULZWNO-UHFFFAOYSA-N
Granular Solid
5g
M.W. 1,000
C25H53NO12
mPEG-amine
559.69
White
Chemical Identifiers
80506-64-5
559.69
MSKSQCLPULZWNO-UHFFFAOYSA-N
C25H53NO12
MFCD00131599
mPEG-amine
Safety and Handling
Safety and Handling
TSCA : TSCA
SDS Documents