missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(Methoxymethyl)diphenylphosphine oxide, 98%, ACROS Organics™

Chemical Identifiers

CAS 4455-77-0
Molecular Formula C14H15O2P
Molecular Weight (g/mol) 246.246
MDL Number MFCD00015451
InChI Key OEPKDBQOTLDTNC-UHFFFAOYSA-N
Synonym methoxymethyl diphenylphosphine oxide, methoxymethyldiphenylphosphine oxide, diphenyl methoxymethyl phosphine oxide, phosphine oxide, methoxymethyl diphenyl, methoxymethyl diphenyl phosphine oxide, phosphine oxide, diphenyl methoxymethyl, methoxymethyl diphenyl phosphine oxide, methoxymethyl phenyl phosphoroso benzene, acmc-1ahy3, cbmicro_008972
PubChem CID 78203
IUPAC Name [methoxymethyl(phenyl)phosphoryl]benzene
SMILES COCP(=O)(C1=CC=CC=C1)C2=CC=CC=C2
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC389690250 View Documents Acros Organics
AC389690250
25g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

For C-C Bond Formation (Olefination)
Specifications

Chemical Identifiers

4455-77-0
246.246
OEPKDBQOTLDTNC-UHFFFAOYSA-N
78203
COCP(=O)(C1=CC=CC=C1)C2=CC=CC=C2
C14H15O2P
MFCD00015451
methoxymethyl diphenylphosphine oxide, methoxymethyldiphenylphosphine oxide, diphenyl methoxymethyl phosphine oxide, phosphine oxide, methoxymethyl diphenyl, methoxymethyl diphenyl phosphine oxide, phosphine oxide, diphenyl methoxymethyl, methoxymethyl diphenyl phosphine oxide, methoxymethyl phenyl phosphoroso benzene, acmc-1ahy3, cbmicro_008972
[methoxymethyl(phenyl)phosphoryl]benzene

Specifications

Authentic
White to Yellow
25g
97.5
98%
MFCD00015451
OEPKDBQOTLDTNC-UHFFFAOYSA-N
[methoxymethyl(phenyl)phosphoryl]benzene
78203
Crystalline Powder
(Methoxymethyl)diphenylphosphine oxide
Glass bottle
114.0°C to 117.0°C
4455-77-0
100.0
C14H15O2P
methoxymethyl diphenylphosphine oxide, methoxymethyldiphenylphosphine oxide, diphenyl methoxymethyl phosphine oxide, phosphine oxide, methoxymethyl diphenyl, methoxymethyl diphenyl phosphine oxide, phosphine oxide, diphenyl methoxymethyl, methoxymethyl diphenyl phosphine oxide, methoxymethyl phenyl phosphoroso benzene, acmc-1ahy3, cbmicro_008972
COCP(=O)(C1=CC=CC=C1)C2=CC=CC=C2
246.246
246.24
98%
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.