missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(Methoxymethyl)diphenylphosphine oxide, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC389690050

 View more versions of this product

Catalog No. AC389690050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

(Methoxymethyl)diphenylphosphine oxide
114° to 117°C
4455-77-0
100.0
Glass bottle
≥97.5%
C14H15O2P
MFCD00015451
White to pale yellow
5g
97.5
Authentic
Crystalline Powder
98%
246.24

Chemical Identifiers

4455-77-0
100.0
C14H15O2P
MFCD00015451
≥97.5%
97.5
98%
246.24
(Methoxymethyl)diphenylphosphine oxide

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.