missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methoxylamine hydrochloride, 25-30% aq. soln.

$80.64 - $80.64

Chemical Identifiers

CAS 593-56-6
Molecular Formula CH3ONH2·HCl
Formula Weight 83.52
MDL Number MFCD00012951
UN Number UN3265
Beilstein 3589723
Merck Index 14,5989
Synonym Methoxyamine hydrochloride; O-Methylhydroxylamine hydrochloride
Chemical Name or Material Methoxylamine hydrochloride
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0841514 SDS Alfa Aesar™
L0841514
25g Each for $80.64
AAL0841522 SDS Alfa Aesar™
L0841522
100g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

Applications
Methoxyamine hydrochloride was used as a reagent in the preparation of O-methyl oximes and also used in the synthesis of O-methyl oximes from aldehydes2 or ketones. It is also applied as an internal standard for Prostaglandin assays by gas chromatography-mass spectrometry.

Solubility
Fully miscible in water. Soluble in DMSO and methanol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Description & Specifications

Chemical Identifiers

593-56-6
83.52
UN3265
14,5989
Methoxylamine hydrochloride
CH3ONH2·HCl
MFCD00012951
3589723
Methoxyamine hydrochloride; O-Methylhydroxylamine hydrochloride

Specifications

25 to 30% aq. Soln.
593-56-6
83.52
UN3265
14,5989
Methoxylamine hydrochloride
25g
CH3ONH2·HCl
MFCD00012951
3589723
Methoxyamine hydrochloride; O-Methylhydroxylamine hydrochloride

Safety and Handling

GHS H Statement
H301-H314-H318-H317
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

RTECSNumber : NC3980000

TSCA : TSCA