Please login to your online account to display your discounted pricing

Alfa Aesar™ Methoxydimethylphenylsilane, 97%

$213.96 - $213.96

Chemical Identifiers

Chemical Name or Material Methoxydimethylphenylsilane
CAS 17881-88-8
Molecular Formula C9H14OSi
Formula Weight 166.3
MDL Number MFCD09266142
Synonym Dimethylphenylmethoxysilane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2716506

 
alfa aesar™
H2716506
5g Each for $213.96
Description & Specifications

Specifications

Formula Weight 166.3
Chemical Name or Material Methoxydimethylphenylsilane
Assay 97%
Molecular Formula C9H14OSi
MDL Number MFCD09266142
CAS 17881-88-8
Synonym Dimethylphenylmethoxysilane
Boiling Point 82°C (28mmHg)
Quantity 5g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.