Please login to your online account to display your discounted pricing

Alfa Aesar™ Methoxydimethyl(p-tolyl)silane, 94%

$62.68 - $183.82

Chemical Identifiers

Chemical Name or Material Methoxydimethyl(p-tolyl)silane
CAS 51501-87-2
Molecular Formula C10H16OSi
Formula Weight 180.32
MDL Number MFCD09266166
Synonym Dimethylmethoxy(4-methylphenyl)silane; Dimethyl(4-methylphenyl)methoxysilane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2794303

  SDS
alfa aesar™
H2794303
1g Each for $62.68

AAH2794306

  SDS
alfa aesar™
H2794306
5g Each for $183.82
Description & Specifications

Specifications

Formula Weight 180.32
Chemical Name or Material Methoxydimethyl(p-tolyl)silane
Assay 94%
Molecular Formula C10H16OSi
MDL Number MFCD09266166
CAS 51501-87-2
Synonym Dimethylmethoxy(4-methylphenyl)silane; Dimethyl(4-methylphenyl)methoxysilane
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.