missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methoxycarbonylsulfenyl chloride, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC308840050

 View more versions of this product

Catalog No. AC308840050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥96.0%
Liquid
Clear bright yellow
CH3O2CSCl
Authentic
97%
MFCD00013648
C2H3ClO2S
126.56
5mL
1.3900g/mL
Methoxycarbonylsulfenyl chloride
Glass bottle
26555-40-8
1.4810 to 1.4830 (20°C, 589nm)

Chemical Identifiers

97%
CH3O2CSCl
MFCD00013648
≥96.0%
C2H3ClO2S
126.56
Methoxycarbonylsulfenyl chloride

Safety and Handling

SDS & Safety Documents

GHS H Statement
Causes severe skin burns and eye damage. Flammable liquid and vapor.

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

Danger