missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methoxycarbonylsulfenyl chloride, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 26555-40-8 |
---|---|
Molecular Formula | C2H3ClO2S |
Molecular Weight (g/mol) | 126.56 |
MDL Number | MFCD00013648 |
InChI Key | TXJXPZVVSLAQOQ-UHFFFAOYSA-N |
Synonym | methoxycarbonylsulfenyl chloride, methoxycarbonyl sulfenyl chloride, s-chloro o-methyl thiocarbonate, thiocarbonic acid, anhydrosulphide with methyl thiohypochlorite, carbonothioic acid, anhydrosulfide with thiohypochlorous acid, methyl ester, acmc-209gs8, methoxycarbonylsulfenylchloride, chlorothio methoxy oxomethane, methoxy carbonylsulfenyl chloride, methoxy-carbonylsulfenyl chloride |
PubChem CID | 123417 |
IUPAC Name | methyl chlorosulfanylformate |
SMILES | COC(=O)SCl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC308840010
|
Thermo Scientific Chemicals
308840010 |
1mL | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC308840050
|
Thermo Scientific Chemicals
308840050 |
5mL | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
26555-40-8 | |
126.56 | |
TXJXPZVVSLAQOQ-UHFFFAOYSA-N | |
123417 | |
COC(=O)SCl |
C2H3ClO2S | |
MFCD00013648 | |
methoxycarbonylsulfenyl chloride, methoxycarbonyl sulfenyl chloride, s-chloro o-methyl thiocarbonate, thiocarbonic acid, anhydrosulphide with methyl thiohypochlorite, carbonothioic acid, anhydrosulfide with thiohypochlorous acid, methyl ester, acmc-209gs8, methoxycarbonylsulfenylchloride, chlorothio methoxy oxomethane, methoxy carbonylsulfenyl chloride, methoxy-carbonylsulfenyl chloride | |
methyl chlorosulfanylformate |
Specifications
26555-40-8 | |
133°C to 134°C | |
Authentic | |
Glass bottle | |
1.4810 to 1.483 | |
MFCD00013648 | |
1.39 | |
TXJXPZVVSLAQOQ-UHFFFAOYSA-N | |
methyl chlorosulfanylformate | |
123417 | |
97% |
1.3900g/mL | |
54°C | |
97% | |
C2H3ClO2S | |
CH3O2CSCl | |
1mL | |
methoxycarbonylsulfenyl chloride, methoxycarbonyl sulfenyl chloride, s-chloro o-methyl thiocarbonate, thiocarbonic acid, anhydrosulphide with methyl thiohypochlorite, carbonothioic acid, anhydrosulfide with thiohypochlorous acid, methyl ester, acmc-209gs8, methoxycarbonylsulfenylchloride, chlorothio methoxy oxomethane, methoxy carbonylsulfenyl chloride, methoxy-carbonylsulfenyl chloride | |
COC(=O)SCl | |
126.56 | |
126.56 | |
Methoxycarbonylsulfenyl chloride |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
EINECSNumber : 247-801-3
RUO â Research Use Only