Please login to your online account to display your discounted pricing

Alfa Aesar™ (Methoxycarbonylmethyl)triphenylphosphonium chloride, 97%

$492.72 - $492.72

Chemical Identifiers

Chemical Name or Material (Methoxycarbonylmethyl)triphenylphosphonium chloride
CAS 2181-97-7
Molecular Formula CH3O2CCH2P(C6H5)3Cl
Formula Weight 370.82
MDL Number MFCD00050199
Beilstein 4172737
Synonym (Carbomethoxymethyl)triphenylphosphonium chloride; Methyl (triphenylphosphonio)acetate chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1777530

  SDS
alfa aesar™
A1777530
250g Each for $492.72
Description & Specifications

Specifications

Formula Weight 370.82
MDL Number MFCD00050199
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point ≈162°C (dec.)
Chemical Name or Material (Methoxycarbonylmethyl)triphenylphosphonium chloride
Assay 97%
Molecular Formula CH3O2CCH2P(C6H5)3Cl
CAS 2181-97-7
Beilstein 4172737
Synonym (Carbomethoxymethyl)triphenylphosphonium chloride; Methyl (triphenylphosphonio)acetate chloride
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 250g

Applications
It is employed as intermediate for pharmaceutical.

Solubility
Very Soluble in water (100 g/L) (25°C).

Notes
Hygroscopic, store away from moisture. Store in a tightly closed container in a cool, dry, well-ventilated area. Store away from oxidizing agent.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.