missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methoxyacetyl chloride, 97%, stabilized, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC125702500

 View more versions of this product

Catalog No. AC125702500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

Methoxyacetyl chloride
28°C
Glass bottle
1.187
112°C to 113°C
250g
38870-89-2
C3H5ClO2
MFCD00000728
2-methoxyacetylchloride, acetyl chloride, methoxy, ch3och2cocl, methoxy acetyl chloride, methoxy acetylchloride, methoxy-acetyl chloride, methoxyacetic acid chloride, methoxyacetyl chloride, methoxyacetylchloride, unii-t66s02b1s3
JJKWHOSQTYYFAE-UHFFFAOYSA-N
2-methoxyacetyl chloride
96623
Liquid
1.1870g/mL
Authentic
1.4190 to 1.422
0.3%mgO
Colorless to Yellow
Stabilized
97%
CH3OCH2COCl
12,310
Solubility in water: reacts
COCC(=O)Cl
108.52
108.52
97%

Chemical Identifiers

38870-89-2
108.52
JJKWHOSQTYYFAE-UHFFFAOYSA-N
96623
COCC(=O)Cl
C3H5ClO2
MFCD00000728
2-methoxyacetylchloride, acetyl chloride, methoxy, ch3och2cocl, methoxy acetyl chloride, methoxy acetylchloride, methoxy-acetyl chloride, methoxyacetic acid chloride, methoxyacetyl chloride, methoxyacetylchloride, unii-t66s02b1s3
2-methoxyacetyl chloride
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 254-169-2

SDS Documents
Product Certifications