missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methoxyacetonitrile, 98%, ACROS Organics™

Chemical Identifiers

CAS 1738-36-9
Molecular Formula C3H5NO
Molecular Weight (g/mol) 71.08
InChI Key QKPVEISEHYYHRH-UHFFFAOYSA-N
Synonym 2-methoxy acetonitrile, 2-methoxy-acetonitrile, acetonitrile, 2-methoxy, acetonitrile, methoxy, acetonitrile,2-methoxy, acmc-209e7f, intermediates-zcf02294, methoxyacetonitrile, ncch2och3
PubChem CID 74442
IUPAC Name 2-methoxyacetonitrile
SMILES COCC#N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC125661000
SDS View Documents
Thermo Fisher Scientific
AC125661000
100g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

1738-36-9
71.08
2-methoxy acetonitrile, 2-methoxy-acetonitrile, acetonitrile, 2-methoxy, acetonitrile, methoxy, acetonitrile,2-methoxy, acmc-209e7f, intermediates-zcf02294, methoxyacetonitrile, ncch2och3
2-methoxyacetonitrile
C3H5NO
QKPVEISEHYYHRH-UHFFFAOYSA-N
74442
COCC#N

Specifications

0.9500g/mL
Authentic
1.3800 to 1.3830 (20°C, 589nm)
0.5% max.
Colorless to Yellow
1738-36-9
C3H5NO
03,242
2-methoxy acetonitrile, 2-methoxy-acetonitrile, acetonitrile, 2-methoxy, acetonitrile, methoxy, acetonitrile,2-methoxy, acmc-209e7f, intermediates-zcf02294, methoxyacetonitrile, ncch2och3
COCC#N
71.08
71.08
98%
31°C
Glass bottle
0.95
118.0°C to 119.0°C (731.0 mmHg)
100g
97.5% min. (GC)
CH3OCH2CN
05,436
QKPVEISEHYYHRH-UHFFFAOYSA-N
2-methoxyacetonitrile
74442
Liquid
Methoxyacetonitrile
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing.
Wash face,hands and any exposed skin thoroughly after handling.
IF ON SKIN: Wash with plenty of soap a

GHS Signal Word: Warning

EINECSNumber : 217-092-5

SDS