missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methoxyacetone 96.0+%, TCI America™
$46.60 - $139.81
Chemical Identifiers
| CAS | 5878-19-3 |
|---|---|
| Molecular Formula | C4H8O2 |
| Molecular Weight (g/mol) | 88.11 |
| MDL Number | MFCD00008768 |
| InChI Key | CUZLJOLBIRPEFB-UHFFFAOYSA-N |
| Synonym | methoxyacetone, methoxy-2-propanone, 2-propanone, 1-methoxy, 1-methoxy-2-propanone, 1-methoxyacetone, methoxyaceton, methoxymethyl methyl ketone, methoxy acetone, ch3coch2och3 |
| PubChem CID | 22172 |
| IUPAC Name | 1-methoxypropan-2-one |
| SMILES | COCC(C)=O |
Chemical Identifiers
| 5878-19-3 | |
| 88.11 | |
| CUZLJOLBIRPEFB-UHFFFAOYSA-N | |
| 22172 | |
| COCC(C)=O |
| C4H8O2 | |
| MFCD00008768 | |
| methoxyacetone, methoxy-2-propanone, 2-propanone, 1-methoxy, 1-methoxy-2-propanone, 1-methoxyacetone, methoxyaceton, methoxymethyl methyl ketone, methoxy acetone, ch3coch2och3 | |
| 1-methoxypropan-2-one |
Specifications
| 5878-19-3 | |
| 115°C | |
| MFCD00008768 | |
| 1993 | |
| CUZLJOLBIRPEFB-UHFFFAOYSA-N | |
| 1-methoxypropan-2-one | |
| 22172 | |
| ≥96.0% (GC) | |
| Methoxyacetone |
| Yellow | |
| C4H8O2 | |
| 1 mL | |
| methoxyacetone, methoxy-2-propanone, 2-propanone, 1-methoxy, 1-methoxy-2-propanone, 1-methoxyacetone, methoxyaceton, methoxymethyl methyl ketone, methoxy acetone, ch3coch2och3 | |
| COCC(C)=O | |
| 88.11 | |
| 88.11 | |
| Liquid |
Safety and Handling
RTECSNumber : UC2988600
TSCA : Yes