missing translation for 'onlineSavingsMsg'
Learn More

Methoxyacetone 96.0+%, TCI America™

Supplier:  TCI America M09061ML

 View more versions of this product

Catalog No. M09061ML

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Methoxyacetone
Yellow
C4H8O2
1 mL
methoxyacetone, methoxy-2-propanone, 2-propanone, 1-methoxy, 1-methoxy-2-propanone, 1-methoxyacetone, methoxyaceton, methoxymethyl methyl ketone, methoxy acetone, ch3coch2och3
COCC(C)=O
88.11
88.11
Liquid
5878-19-3
115°C
MFCD00008768
1993
CUZLJOLBIRPEFB-UHFFFAOYSA-N
1-methoxypropan-2-one
22172
≥96.0% (GC)

Chemical Identifiers

5878-19-3
88.11
CUZLJOLBIRPEFB-UHFFFAOYSA-N
22172
COCC(C)=O
C4H8O2
MFCD00008768
methoxyacetone, methoxy-2-propanone, 2-propanone, 1-methoxy, 1-methoxy-2-propanone, 1-methoxyacetone, methoxyaceton, methoxymethyl methyl ketone, methoxy acetone, ch3coch2och3
1-methoxypropan-2-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : UC2988600

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions