Learn More
Methoxyacetic anhydride, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6454122
Description
Methoxyacetic anhydride is used as the intermediate of 5-Aza-2'-deoxycytidine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethoxyacetic anhydride is used as the intermediate of 5-Aza-2′-deoxycytidine.
Solubility
Very Soluble in water.(193 g/L) (25°C)
Notes
Moisture sensitive. Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Specifications
Methoxyacetic anhydride | |
Colorless | |
(CH3OCH2CO)2O | |
100 g | |
1771552 | |
methoxyacetic anhydride, methoxyacetic acid anhydride, acetic acid, methoxy-, anhydride, 2-methoxyacetic anhydride, acetic acid,2-methoxy-, anhydride, di methoxyacetic anhydride, methoxy acetic acid anhydride, methoxy-acetic acid anhydride, 2-methoxyacetyl 2-methoxyacetate, methoxyacetic anhydride?intermediate of 5-aza-2'-deoxycytidine | |
PEHFQQWAINXOQG-UHFFFAOYSA-N | |
(2-methoxyacetyl) 2-methoxyacetate | |
88095 | |
95% |
19500-95-9 | |
C6H10O5 | |
MFCD11976032 | |
UN3265 | |
Moisture sensitive | |
Very Soluble in water.(193g/L) (25°C) | |
COCC(=O)OC(=O)COC | |
162.14 | |
162.14 | |
Liquid |
Chemical Identifiers
19500-95-9 | |
162.14 | |
PEHFQQWAINXOQG-UHFFFAOYSA-N | |
88095 | |
COCC(=O)OC(=O)COC |
C6H10O5 | |
MFCD11976032 | |
methoxyacetic anhydride, methoxyacetic acid anhydride, acetic acid, methoxy-, anhydride, 2-methoxyacetic anhydride, acetic acid,2-methoxy-, anhydride, di methoxyacetic anhydride, methoxy acetic acid anhydride, methoxy-acetic acid anhydride, 2-methoxyacetyl 2-methoxyacetate, methoxyacetic anhydride?intermediate of 5-aza-2'-deoxycytidine | |
(2-methoxyacetyl) 2-methoxyacetate |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 243-114-8
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only