missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methoxyacetic anhydride, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6454114

 View more versions of this product

Catalog No. AAH6454114



Description

Description

Methoxyacetic anhydride is used as the intermediate of 5-Aza-2'-deoxycytidine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methoxyacetic anhydride is used as the intermediate of 5-Aza-2′-deoxycytidine.

Solubility
Very Soluble in water.(193 g/L) (25°C)

Notes
Moisture sensitive. Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Specifications

Specifications

Methoxyacetic anhydride
Colorless
(CH3OCH2CO)2O
25 g
1771552
methoxyacetic anhydride, methoxyacetic acid anhydride, acetic acid, methoxy-, anhydride, 2-methoxyacetic anhydride, acetic acid,2-methoxy-, anhydride, di methoxyacetic anhydride, methoxy acetic acid anhydride, methoxy-acetic acid anhydride, 2-methoxyacetyl 2-methoxyacetate, methoxyacetic anhydride?intermediate of 5-aza-2'-deoxycytidine
PEHFQQWAINXOQG-UHFFFAOYSA-N
(2-methoxyacetyl) 2-methoxyacetate
88095
95%
19500-95-9
C6H10O5
MFCD11976032
UN3265
Moisture sensitive
Very Soluble in water.(193g/L) (25°C)
COCC(=O)OC(=O)COC
162.14
162.14
Liquid

Chemical Identifiers

19500-95-9
162.14
PEHFQQWAINXOQG-UHFFFAOYSA-N
88095
COCC(=O)OC(=O)COC
C6H10O5
MFCD11976032
methoxyacetic anhydride, methoxyacetic acid anhydride, acetic acid, methoxy-, anhydride, 2-methoxyacetic anhydride, acetic acid,2-methoxy-, anhydride, di methoxyacetic anhydride, methoxy acetic acid anhydride, methoxy-acetic acid anhydride, 2-methoxyacetyl 2-methoxyacetate, methoxyacetic anhydride?intermediate of 5-aza-2'-deoxycytidine
(2-methoxyacetyl) 2-methoxyacetate
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 243-114-8

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only