missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methoxyacetic acid hydrazide, 95%

$153.12 - $607.59

Chemical Identifiers

CAS 20605-41-8
Molecular Formula C3H8N2O2
Molecular Weight (g/mol) 104.109
MDL Number MFCD02197887
InChI Key XEPXDMNZXBUSOI-UHFFFAOYSA-N
Synonym methoxyacetic acid hydrazide, methoxyacetohydrazide, acetic acid, methoxy-, hydrazide, 2-methoxyethanehydrazide, 2-methoxyacetic acid hydrazide, acetic acid,2-methoxy-, hydrazide, methoxymethyl-carboxylic acid hydrazide
PubChem CID 2759532
IUPAC Name 2-methoxyacetohydrazide
SMILES COCC(=O)NN
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6557306 View Documents Alfa Aesar™
H6557306
5g Each for $153.12
 
AAH6557314 View Documents Alfa Aesar™
H6557314
25g Each for $607.59
 
Specifications

Chemical Identifiers

20605-41-8
104.109
XEPXDMNZXBUSOI-UHFFFAOYSA-N
2759532
COCC(=O)NN
C3H8N2O2
MFCD02197887
methoxyacetic acid hydrazide, methoxyacetohydrazide, acetic acid, methoxy-, hydrazide, 2-methoxyethanehydrazide, 2-methoxyacetic acid hydrazide, acetic acid,2-methoxy-, hydrazide, methoxymethyl-carboxylic acid hydrazide
2-methoxyacetohydrazide

Specifications

5g
C3H8N2O2
MFCD02197887
methoxyacetic acid hydrazide, methoxyacetohydrazide, acetic acid, methoxy-, hydrazide, 2-methoxyethanehydrazide, 2-methoxyacetic acid hydrazide, acetic acid,2-methoxy-, hydrazide, methoxymethyl-carboxylic acid hydrazide
COCC(=O)NN
104.109
104.11
95%
20605-41-8
CH3OCH2CONHNH2
1903182
XEPXDMNZXBUSOI-UHFFFAOYSA-N
2-methoxyacetohydrazide
2759532
Solid
Methoxyacetic acid hydrazide