Learn More
Methoxyacetic acid, 97+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 186770010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Methoxyacetic acid | |
625-45-6 | |
Colorless to Yellow | |
202°C to 204°C | |
Authentic | |
Glass bottle | |
1.4153 to 1.4173 | |
MFCD00004308 | |
03, 232 | |
methoxyacetic acid, acetic acid, methoxy, methoxyethanoic acid, acetic acid, 2-methoxy, ch3och2cooh, methoxyacetlc acid, unii-f11t1h7q7w, methoxy acetic acid, methoxy-acetic acid, ccris 6518 | |
RMIODHQZRUFFFF-UHFFFAOYSA-N | |
2-methoxyacetic acid | |
12251 | |
97+% |
98% | |
7°C | |
1.1740g/mL | |
121°C | |
97+% | |
C3H6O3 | |
CH3OCH2CO2H | |
1 L | |
1.174 | |
Solubility in water: freely soluble | |
COCC(=O)O | |
90.08 | |
90.08 | |
Liquid |
Chemical Identifiers
625-45-6 | |
90.08 | |
RMIODHQZRUFFFF-UHFFFAOYSA-N | |
12251 | |
COCC(=O)O |
C3H6O3 | |
MFCD00004308 | |
methoxyacetic acid, acetic acid, methoxy, methoxyethanoic acid, acetic acid, 2-methoxy, ch3och2cooh, methoxyacetlc acid, unii-f11t1h7q7w, methoxy acetic acid, methoxy-acetic acid, ccris 6518 | |
2-methoxyacetic acid |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes severe skin burns and eye damage.
Harmful if swallowed.
May damage fertility.
May damage the unborn child.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
EINECSNumber : 210-894-6
RTECSNumber : AI8650000
TSCA : TSCA
RUO – Research Use Only