Please login to your online account to display your discounted pricing

Methoxy(cyclooctadiene)iridium(I) dimer, Ir nominally 58%, Alfa Aesar™

Light, Air & Moisture Sensitive

Chemical Identifiers

Chemical Name or Material Methoxy(cyclooctadiene)iridium(I) dimer
CAS 12148-71-9
Physical Form Powder
Assay Percent Range Ir nominally 58%
Molecular Formula C18H30Ir2O2
Formula Weight 662.88
MDL Number MFCD08459360
Synonym 1, 5-Cyclooctadiene(methoxy)iridium(I) dimer
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4496703

  SDS
alfa aesar
4496703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4496706

  SDS
alfa aesar
4496706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Formula Weight 662.88
Quantity 1g
Color Yellow
Chemical Name or Material Methoxy(cyclooctadiene)iridium(I) dimer
Composition Ir nominally 58%
Assay Percent Range Ir nominally 58%
Molecular Formula C18H30Ir2O2
MDL Number MFCD08459360
CAS 12148-71-9
Synonym 1, 5-Cyclooctadiene(methoxy)iridium(I) dimer
Sensitivity Light, air, moisture sensitive
Health Hazard 1 H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Health Hazard 2 In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves.
Safety and Handling

H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves.