missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methoxy(cyclooctadiene)iridium(I) dimer, Ir nominally 58%, Alfa Aesar™

Light, Air & Moisture Sensitive

Manufacturer:  Alfa Aesar 4496706

 View more versions of this product

Catalog No. AA4496706



Specifications

Specifications

Methoxy(cyclooctadiene)iridium(I) dimer
Yellow
12148-71-9
C18H32Ir2O2
bis 1,5-cyclooctadiene di-mu-methoxydiiridium i, 1,5-cyclooctadiene methoxy iridium i dimer
MNKCGUKVRJZKEQ-MIXQCLKLSA-N
(1Z,5Z)-cycloocta-1,5-diene;iridium;methanol
124202702
Powder
Light, air, moisture sensitive
5g
Ir nominally 58%
MFCD08459360
Ir nominally 58%
CO.CO.C1CC=CCCC=C1.C1CC=CCCC=C1.[Ir].[Ir]
664.886
662.88

Chemical Identifiers

12148-71-9
664.886
MNKCGUKVRJZKEQ-MIXQCLKLSA-N
124202702
CO.CO.C1CC=CCCC=C1.C1CC=CCCC=C1.[Ir].[Ir]
C18H32Ir2O2
MFCD08459360
bis 1,5-cyclooctadiene di-mu-methoxydiiridium i, 1,5-cyclooctadiene methoxy iridium i dimer
(1Z,5Z)-cycloocta-1,5-diene;iridium;methanol
Safety and Handling

Safety and Handling

In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves.

H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

SDS Documents