Please login to your online account to display your discounted pricing

Alfa Aesar™ Methotrexate disodium salt, 98.5%

Inhibits the metabolism of folic acid

$153.69 - $153.69

Chemical Identifiers

Chemical Name or Material Methotrexate disodium salt
CAS 7413-34-5
Formula Weight 498.4
MDL Number MFCD00274560
UN Number UN2811
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66364MD

  SDS
alfa aesar™
J66364MD
250mg Each for $153.69
Description & Specifications

Specifications

Formula Weight 498.4
Chemical Name or Material Methotrexate disodium salt
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Physical Form Powder
Assay 98.5%
MDL Number MFCD00274560
CAS 7413-34-5
Boiling Point 823°C
DOT Information Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Color Yellow
Health Hazard 1 Danger
Quantity 250mg
Sensitivity Hygroscopic
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number MA1232000
UN Number UN2811
Solubility Information Soluble in water
Storage Note 1 2° to 8°C
Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.

2° to 8°C