Please login to your online account to display your discounted pricing

Alfa Aesar™ Methoprene acid, 98%


Quantity 5mg

Manufacturer: alfa aesar™  J67031LB0

 View more versions of this product

Catalog No. AAJ67031LB0

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 268.4
Chemical Name or Material Methoprene acid
Assay 98%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym 11-Methoxy-3,7,11-trimethyl-2E,4E-dodecadienoic acid
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD01317817
Quantity 5mg
TSCA TSCA
Health Hazard 1 Warning
Physical Form Oil
Color Pale yellow
Storage Note 1 -30° to -10°C
CAS 53092-52-7

Specifications

Chemical Name or Material Methoprene acid
CAS 53092-52-7
Formula Weight 268.4
MDL Number MFCD01317817
Synonym 11-Methoxy-3,7,11-trimethyl-2E,4E-dodecadienoic acid
Accessories

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

-30° to -10°C

SDS & Safety Documents