Please login to your online account to display your discounted pricing

Method 552.2 Methyl Esters, SPEX CertiPrep™

$266.60 - $266.60

Chemical Identifiers

Chemical Name or Material Methyl Esters Mix
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

5522MEO

 
spex certiprep llc
5522-MEO
2mL amber prescored glass ampule 1mL Each for $266.60
Description & Specifications

Specifications

Quantity 1mL
Chemical Name or Material Methyl Esters Mix
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Packaging 2mL amber prescored glass ampule
Recommended Storage Freezer
Methods For US EPA Methods 552, 552.2
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
DOT Information Nonhazardous
Solvent or Matrix MTBE
Analyte or Component Names 400μg/mL Dalapon methyl ester; 600μg/mL Methyl monochloroacetate; 400μg/mL Methyl mono-bromacetate; 400μg/mL Methyl bromo-chloroacetate; 400μg/mL Methyl bromodi-chlroacetate; 1000μg/mL Methyl chlorodi-bromoacetate; 200μg/mL Methyl dibromoacetate; 1000μg/mL Methyl 2,3-dibromopropionate (surrogate); 600μg/mL Methyl dichloroacetate; 2000μg/mL Methyl tribromoacetate; 200μg/mL Methyl trichloroacetate

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

Freezer