Please login to your online account to display your discounted pricing

Methimepip dihydrobromide, Tocris

Extremely selective H3 agonist

$218.74 - $905.98

Chemical Identifiers

Chemical Name or Material Methimepip dihydrobromide
Assay Percent Range >98%
Molecular Formula C10H17N3·2HBr
Formula Weight 341.09
Synonym 4-(1H-Imidazol-4-ylmethyl)-1-methylpiperidine dihydrobromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

294710

 
r & d systems
RDS294710
10mg Each for $218.74

294750

 
r & d systems
RDS294750
50mg Each for $905.98
Description & Specifications

Specifications

Formula Weight 341.09
Quantity 10mg
Assay Percent Range >98%
Chemical Name or Material Methimepip dihydrobromide
Molecular Formula C10H17N3·2HBr
CAS 817636-54-7
Synonym 4-(1H-Imidazol-4-ylmethyl)-1-methylpiperidine dihydrobromide
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature