Please login to your online account to display your discounted pricing

Alfa Aesar™ Methicillin sodium salt, 98%

Chemical Identifiers

Chemical Name or Material Methicillin sodium salt
CAS 132-92-3
Formula Weight 402.4
MDL Number MFCD07787409
Merck Index 14,5969
Synonym Sodium (2,6-dimethoxyphenyl)penicillin; Sodium methicillin
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ6641403

  SDS
alfa aesar™
J6641403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 196° to 197°C (dec.)
Formula Weight 402.4
Health Hazard 3 GHS P Statement
P285-P261-P280-P305+P351+P338-P405-P501a
In case of inadequate ventilation wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H334-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Chemical Name or Material Methicillin sodium salt
CAS 132-92-3
Physical Form Powder
Assay 98%
MDL Number MFCD07787409
Merck Index 14,5969
Boiling Point 640°C
Solubility Information Slightly soluble in acetone; Soluble in ethanol or water
Sensitivity Light Sensitive
Quantity 1g
Synonym Sodium (2,6-dimethoxyphenyl)penicillin; Sodium methicillin
Health Hazard 1 Danger
RTECS Number XH8950000
Color White to off-white
Storage Note 1 -30° to -10°C

Applications
Targets primarily the cell wall of Gram-positive organisms especially Staphylococcus aureus. Methicillin is used to study bacterium susceptibility, and to inhibit cell-wall synthesis. PBP′s are enzymes which catalyze a pentaglycine crosslink between alanine and lysine residues providing additional strength to the cell wall. Without a pentaglycine crosslink, the integrity of the cell wall is severely compromised and ultimately leads to cell lysis and death. Methicillin Sodium Salt is inactivated by gastric acid so it is given to cells by other means in which gastric acid is not present in vitro.

Solubility
Slightly soluble in acetone. Soluble in ethanol or water

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.

Safety and Handling

Danger

GHS P Statement
P285-P261-P280-P305+P351+P338-P405-P501a
In case of inadequate ventilation wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H334-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

-30° to -10°C