Please login to your online account to display your discounted pricing

Alfa Aesar™ Methicillin sodium salt, 50mg/mL in distilled water, sterile-filtered

Chemical Identifiers

Chemical Name or Material Methicillin sodium salt
CAS 132-92-3
Name Note 50mg/mL in distilled water, sterile-filtered
MDL Number MFCD07787409
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ671008EQ

 
alfa aesar™
J671008EQ
5 x 1mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67100XF

 
alfa aesar™
J67100XF
1mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Methicillin sodium salt
CAS 132-92-3
MDL Number MFCD07787409
Physical Form Liquid
Name Note 50mg/mL in distilled water, sterile-filtered
Storage Note 1 -30° to -10°C
Quantity 5 x 1mL
RTECS Number XH8950000
Safety and Handling

-30° to -10°C