missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methicillin sodium salt, 50mg/mL in distilled water, sterile-filtered, Thermo Scientific™
Supplier: Thermo Scientific Chemicals J67100XF
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Methicillin Sodium Salt | |
50mg/mL in distilled water, sterile-filtered | |
132-92-3 | |
MFCD07787409 | |
[Na+].COC1=CC=CC(OC)=C1C(=O)N[C@H]1[C@H]2SC(C)(C)[C@@H](N2C1=O)C([O-])=O | |
402.40 | |
CHEBI:52065 |
Methicillin sodium salt | |
Liquid | |
C17H19N2NaO6S | |
MGFZNWDWOKASQZ-UMLIZJHQSA-M | |
sodium (2S,5R,6R)-6-(2,6-dimethoxybenzamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate | |
23667627 | |
1 mL |
Chemical Identifiers
132-92-3 | |
402.40 | |
MGFZNWDWOKASQZ-UMLIZJHQSA-M | |
CHEBI:52065 | |
[Na+].COC1=CC=CC(OC)=C1C(=O)N[C@H]1[C@H]2SC(C)(C)[C@@H](N2C1=O)C([O-])=O |
C17H19N2NaO6S | |
MFCD07787409 | |
23667627 | |
sodium (2S,5R,6R)-6-(2,6-dimethoxybenzamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate |
Safety and Handling
P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c
H317-H334-H335
EINECSNumber : 205-083-9
RTECSNumber : XH8950000
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C