missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methanol, ultrapure, Spectrophotometric Grade, >99.8%, Alfa Aesar™

Manufacturer:  Alfa Aesar 32435K2

 View more versions of this product

Catalog No. AA32435K2



Specifications

Specifications

Methanol
Sprectrophotometric
>99.8%
MFCD00004595
OKKJLVBELUTLKV-UHFFFAOYSA-N
methanol
887
32.04
Mild
Colorless
67-56-1
Colorless liquid
CH4O
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
CO
32.042
CHEBI:17790
Liquid
128 hPa (96mm Hg) at 20°C
1L

Chemical Identifiers

67-56-1
32.042
OKKJLVBELUTLKV-UHFFFAOYSA-N
887
methanol
CH4O
MFCD00004595
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
CHEBI:17790
CO
Safety and Handling

Safety and Handling

H225-H301-H311-H331-H370: Highly flammable liquid and vapour. Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. Causes damage to organs.

DOTInformation : DOT Class: 3; Packing Group: II

EINECSNumber : 200-659-6

SDS Documents