Please login to your online account to display your discounted pricing

Methanol, ultrapure, Spectrophotometric Grade, 99.8+%, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Methanol
CAS 67-56-1
Name Note ultrapure
Purity Grade Spectrophotometric grade
Molecular Formula CH3OH
Formula Weight 32.04
MDL Number MFCD00004595
UN Number UN1230
Beilstein 1098229
Merck Index 14,5957
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA32435M6

  SDS
alfa aesar
32435M6
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 32.04
Purity Grade Spectrophotometric grade
Quantity 4x4L
Refractive Index 1.3290
UN Number UN1230
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: METHANOL
Flash Point 11°C (52°F)
Physical Form Liquid
RTECS Number PC1400000
Molecular Formula CH3OH
Health Hazard 1 Danger
Density 0.791
Odor Mild
Beilstein 1098229
TSCA TSCA
Name Note ultrapure
Melting Point −98°C
Boiling Point 64.7°C
CAS 67-56-1
Health Hazard 2 GHS H Statement
H225-H301-H311-H331-H370;
Highly flammable liquid and vapor.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Health Hazard 3 GHS P Statement
P210-P301+P310-P303+P361+P353-P361-P405-P501a;
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower. Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00004595
Assay ≥99.8%
Merck Index 14,5957
Chemical Name or Material Methanol
Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P303+P361+P353-P361-P405-P501a;
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower. Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H301-H311-H331-H370;
Highly flammable liquid and vapor.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.