missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methanol, ultrapure, Spectrophotometric Grade, >99.8%, Alfa Aesar™

$61.74 - $379.53

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.042
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym carbinol, colonial spirit, columbian spirit, methyl alcohol, methyl hydroxide, methylol, pyroxylic spirit, wood alcohol, wood naphtha, wood spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA32435K2 View Documents Thermo Fisher Scientific
32435K2
1L Each for $61.74
 
AA32435K7 SDS View Documents Thermo Fisher Scientific
32435K7
4L Each for $113.53
 
AA32435M6 View Documents Thermo Fisher Scientific
32435M6
4 x 4L Each for $379.53
 
Specifications

Chemical Identifiers

67-56-1
32.042
OKKJLVBELUTLKV-UHFFFAOYSA-N
887
methanol
CH4O
MFCD00004595
carbinol, colonial spirit, columbian spirit, methyl alcohol, methyl hydroxide, methylol, pyroxylic spirit, wood alcohol, wood naphtha, wood spirit
CHEBI:17790
CO

Specifications

67-56-1
>99.8%
MFCD00004595
carbinol, colonial spirit, columbian spirit, methyl alcohol, methyl hydroxide, methylol, pyroxylic spirit, wood alcohol, wood naphtha, wood spirit
OKKJLVBELUTLKV-UHFFFAOYSA-N
methanol
887
32.04
Spectrophotometric
Mild
128 hPa (96mm Hg) at 20°C
64.7°C
°C to 98°C
Methanol
Colorless liquid
CH4O
1098229
Miscible with water and most of the organic solvents including ethyl acetate,toluene,dichloromethane,chloroform,tetrahydrofuran,and acetone.
CO
32.042
CHEBI:17790
Liquid
0.791
1.329
11°C (52°F)
Colorless
1L
Safety and Handling

Safety and Handling

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P307+P311-P311-P312-P330-P363-P370+P378q-P501c

H225-H301+H311+H331-H370-H372

DOTInformation : DOT Class: 3; Packing Group: II

EINECSNumber : 200-659-6

RTECSNumber : PC1400000

TSCA : Yes

Recommended Storage : Ambient temperatures