Learn More
Methanol, Semiconductor Grade, 99.9% min, Thermo Scientific Chemicals
Maximum level of impurities: Color (APHA) 10, Acidity 0.3meq/g, Alkalinity 0.1meq/g, Evaporation residue 5ppm, H2O (by Karl Fischer titrn) 0.05%, Solubility in H2O P.T., Cl 0.2ppm, PO4 0.5ppm
Supplier: Thermo Scientific Chemicals 019393K2
Description
Methanol is widely used as a solvent in synthetic organic chemistry. It is also used as a solvent in analytical chemistry, especially for HPLC, LC-MS, GC-MS, ICP-MS and spectrophotometry. It is used in the production of biodiesel, formaldehyde and acetic acid. It is also used as fuel, antifreeze, denaturant for ethanol and an energy carrier for fuel cell application.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Methanol | |
67-56-1 | |
Colorless liquid | |
64.7°C | |
11°C (52°F) | |
Liquid | |
99.9% min. | |
MFCD00004595 | |
1098229 | |
Miscible with water and most of the organic solvents including ethyl acetate,toluene,dichloromethane,chloroform,tetrahydrofuran,and acetone. | |
CO | |
32.04 | |
CHEBI:17790 | |
Semiconductor |
°C to 98°C | |
Colorless | |
0.791 | |
1 L | |
455°C (851°F) | |
1.329 | |
CH4O | |
128 hPa (96mm Hg) at 20°C | |
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
methanol | |
887 | |
32.04 |
Chemical Identifiers
67-56-1 | |
32.04 | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
887 | |
methanol |
CH4O | |
MFCD00004595 | |
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
CHEBI:17790 | |
CO |
Safety and Handling
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P307+P311-P311-P312-P330-P363-P370+P378q-P501c
H225-H301+H311+H331-H370-H372
DOTInformation : Hazard Class: 3, Packing Group: II
EINECSNumber : 200-659-6
RTECSNumber : PC1400000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only