missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methanol, Semiconductor Grade, 99.9% min., Alfa Aesar™

Maximum level of impurities: Color (APHA) 10, Acidity 0.3meq/g, Alkalinity 0.1meq/g, Evaporation residue 5ppm, H2O (by Karl Fischer titrn) 0.05%, Solubility in H2O P.T., Cl 0.2ppm, PO4 0.5ppm

Manufacturer:  Alfa Aesar 19393AE

 View more versions of this product

Catalog No. AA19393AE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Methanol
Semiconductor Grade
99.9% min.
MFCD00004595
CO
32.042
CHEBI:17790
Liquid
455°C (851°F)
Colorless
67-56-1
Colorless liquid
CH4O
OKKJLVBELUTLKV-UHFFFAOYSA-N
methanol
887
32.04
0.791g/cm3
128 hPa (96mm Hg) at 20°C
100mL

Chemical Identifiers

67-56-1
32.042
OKKJLVBELUTLKV-UHFFFAOYSA-N
CHEBI:17790
CO
CH4O
MFCD00004595
887
methanol
Safety and Handling

Safety and Handling

H225-H301-H311-H331-H370: Highly flammable liquid and vapour. Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. Causes damage to organs.

DOTInformation : Hazard Class: 3, Packing Group: II

EINECSNumber : 200-659-6

SDS Documents