Please login to your online account to display your discounted pricing

Methanol, Semiconductor Grade, 99.9% min., Alfa Aesar™

Maximum level of impurities: Color (APHA) 10, Acidity 0.3meq/g, Alkalinity 0.1meq/g, Evaporation residue 5ppm, H2O (by Karl Fischer titrn) 0.05%, Solubility in H2O P.T., Cl 0.2ppm, PO4 0.5ppm

$137.26 - $137.26

Chemical Identifiers

Chemical Name or Material Methanol
CAS 67-56-1
Purity Grade Semiconductor Grade
Physical Form Liquid
Assay Percent Range 99.9% min.
Molecular Formula CH3OH
Formula Weight 32.04
MDL Number MFCD00004595
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA19393AE

  SDS
alfa aesar
19393AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA19393K2

  SDS
alfa aesar
19393K2
1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA19393K7

  SDS
alfa aesar
19393K7
4L Each for $137.26

AA19393M1

  SDS
alfa aesar
19393M1
4 x 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Formula Weight 32.04
Quantity 100mL
DOT Information Hazard Class: 3, Packing Group: II
Chemical Name or Material Methanol
EINECS Number 200-659-6
Purity Grade Semiconductor Grade
Appearance Colorless liquid
Molecular Formula CH3OH
Health Hazard 1 H225-H301-H311-H331-H370: Highly flammable liquid and vapour. Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. Causes damage to organs.
Density 0.791g/cm3
Ignition Point 455°C (851°F)
Vapor Pressure 128 hPa (96mm Hg) at 20°C
Assay Percent Range 99.9% min.
MDL Number MFCD00004595
CAS 67-56-1
Safety and Handling

H225-H301-H311-H331-H370: Highly flammable liquid and vapour. Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. Causes damage to organs.