missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methanol, Semiconductor Grade, 99.9% min., Alfa Aesar™

Maximum level of impurities: Color (APHA) 10, Acidity 0.3meq/g, Alkalinity 0.1meq/g, Evaporation residue 5ppm, H2O (by Karl Fischer titrn) 0.05%, Solubility in H2O P.T., Cl 0.2ppm, PO4 0.5ppm

Manufacturer:  Alfa Aesar 19393M1

 View more versions of this product

Catalog No. AA19393M1



Specifications

Specifications

Methanol
Colorless liquid
CH4O
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
CO
32.042
CHEBI:17790
Liquid
0.791g/cm3
128 hPa (96mm Hg) at 20°C
4 x 1L
67-56-1
99.9% min.
MFCD00004595
OKKJLVBELUTLKV-UHFFFAOYSA-N
methanol
887
32.04
Semiconductor
455°C (851°F)
Colorless

Chemical Identifiers

67-56-1
32.042
OKKJLVBELUTLKV-UHFFFAOYSA-N
887
methanol
CH4O
MFCD00004595
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
CHEBI:17790
CO
Safety and Handling

Safety and Handling

H225-H301-H311-H331-H370: Highly flammable liquid and vapour. Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. Causes damage to organs.

DOTInformation : Hazard Class: 3, Packing Group: II

EINECSNumber : 200-659-6

SDS Documents