Please login to your online account to display your discounted pricing

Methanol, OmniSolv™, Pesticide Residue Grade, EMD Millipore


Quantity 1L
Packaging Glass Bottle

Manufacturer: emd millipore corporation  MX04846

 View more versions of this product

Catalog No. MMX04846

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Liquid
Formula Weight 32.05
Quantity 1L
Color Clear colorless
Density 0.792g/cm3 (20°C)
Molecular Formula CH4O
Linear Formula CH3OH
Synonym Methyl alcohol
Chemical Name or Material Methanol
Ignition Point 464°C / 867.2°F
Name Note For Pesticide Residue Grade; also for Gas Chromatography
Packaging Glass Bottle
Recommended Storage Room Temperature
Vapor Pressure 12.9 kPa (20°C)
Evaporation Number 2.1
CAS 67-56-1
Explosion Limit 6 to 36.5%

Specifications

Chemical Name or Material Methanol
Name Note For Pesticide Residue Grade; also for Gas Chromatography
CAS 67-56-1
Physical Form Liquid
Molecular Formula CH4O
Linear Formula CH3OH
Formula Weight 32.05
Synonym Methyl alcohol
Accessories

Room Temperature