Please login to your online account to display your discounted pricing

Methanol, OmniSolv™, EMD Millipore

For Purge and Trap Analysis

$305.14 - $305.14

Chemical Identifiers

Chemical Name or Material Methanol
CAS 67-56-1
Molecular Formula CH4O
Formula Weight 32.04
UN Number 1230
Synonym Methyl Alcohol
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MMX04826

 
emd millipore corporation
MX04826
Glass Bottle 1L Each for $305.14
Description & Specifications

Specifications

Boiling Point 64.5°C
Melting Point −97.8°C
Assay ≥99.9%
Formula Weight 32.04
Quantity 1L
Storage Information Store in accordance with local regulations. Store in a segregated and approved area. Store in original container, protected from direct sunlight. Eliminate all ignition sources. Separate from oxidizing materials. Keep container tightly closed and sealed until ready for use. Containers that have been opened must be carefully resealed and kept upright to prevent leakage.
Chemical Name or Material Methanol
Ignition Point 464°C (867.2°F)
Odor Alcohol-like
Packaging Glass Bottle
Vapor Pressure 12.9 kPa (97mm Hg) [20°C]
Flash Point Closed cup: 12°C (53.6°F), Open cup: 15.85°C (60.5°F)
Physical Form Liquid
Color Colorless
Molecular Formula CH4O
CAS 67-56-1
UN Number 1230
Synonym Methyl Alcohol
Solubility Information Soluble in water
Density 0.79g/cm3 at 20°C