Please login to your online account to display your discounted pricing

Methanol, Low water, BakerDRY™, J.T.Baker™

Specifically manufactured for use in organometallic and oligonucleotide synthesis

$150.52 - $278.41

Chemical Identifiers

Chemical Name or Material Methanol
CAS 67-56-1
Name Note Low water
Purity Grade BakerDRY™
Molecular Formula CH3OH
Formula Weight 32.04
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003362

 
avantor performance
909710
Glass Bottle 100mL Each for $278.41

02003363

 
avantor performance
909712
Narrow Mouth Amber Glass Bottle, w/ Septum 1L Each for $150.52
Description & Specifications

Specifications

Formula Weight 32.04
Chemical Name or Material Methanol
Molecular Formula CH3OH
CAS 67-56-1
Packaging Glass Bottle
Specific Gravity 1L = 0.80kg
Name Note Low water
Purity Grade BakerDRY™
Flash Point 12.00°C
Quantity 100mL