missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methanol, LC-MS Grade, ≥99.8%

Solvent for LC-MS analysis

Manufacturer:  Alfa Aesar™ 47192K7

 View more versions of this product

Catalog No. AA47192K7



Specifications

Specifications

Methanol
CH4O
MFCD00004595
1098229
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
CO
32.042
CHEBI:17790
Liquid
LC-MS
Mild
64.7°C
-98°C
67-56-1
CH3OH
UN1230
14,5957
OKKJLVBELUTLKV-UHFFFAOYSA-N
methanol
887
32.04
≥99.8%
0.791g/mL
1.3290
Colorless
4L

Chemical Identifiers

67-56-1
32.042
OKKJLVBELUTLKV-UHFFFAOYSA-N
887
methanol
CH4O
MFCD00004595
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
CHEBI:17790
CO
Safety and Handling

Safety and Handling

GHS H Statement
H225-H301-H311-H331-H370
Highly flammable liquid and vapor.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.

GHS P Statement
P210-P301+P310-P303+P361+P353-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: METHANOL

RTECSNumber : PC1400000

TSCA : TSCA

SDS Documents