missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methanol, LC-MS CHROMASOLV™, Honeywell Riedel-de Haën™
CHROMASOLV™ LC-MS
Supplier: Honeywell Chemicals 349666X1L
Description
- For LC-MS and LC-UV trace metals analysis
- HPLC / UV-VIS quality control
- Low gradient baseline drift
- Low levels of trace metals
- No non-volatile impurities
Specifications
Methanol | |
0.791g/mL at 25°C | |
0.36 | |
725°F | |
CH3OH | |
410 mmHg (50°C), 97.68 mmHg (20°C) | |
1098229 | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
methanol | |
887 | |
32.04g/mol | |
LC-MS CHROMASOLV™ |
67-56-1 | |
6 x 1 L | |
9°C (48.2°F) | |
CH4O | |
MFCD00004595 | |
UN1230 | |
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
CO | |
32.04 | |
CHEBI:17790 | |
≥99.9% |
Chemical Identifiers
67-56-1 | |
32.04 | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
887 | |
methanol |
CH4O | |
MFCD00004595 | |
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
CHEBI:17790 | |
CO |
Safety and Handling
P210-P260-P280-P301 + P310-P311
RTECSNumber : PC1400000