missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methanol, HPLC Grade, Ricca Chemical
Supplier: Ricca Chemical Company R48280004C
Specifications
Methanol | |
67-56-1 | |
100.0 | |
4 L | |
Liquid | |
max. 0.0003 | |
max. 0.0002 | |
max. 0.1 | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
methanol | |
887 | |
min. 99.9% |
for Anethole Blank | |
98.0 | |
Colorless Liquid | |
Amber Glass Bottle | |
1e-05 | |
CH4O | |
MFCD00004595 | |
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
CO | |
32.04 | |
CHEBI:17790 | |
HPLC |
Chemical Identifiers
67-56-1 | |
32.04 | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
887 | |
methanol |
CH4O | |
MFCD00004595 | |
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
CHEBI:17790 | |
CO |