Please login to your online account to display your discounted pricing

Methanol, HPLC Grade, for Anethole Blank, Ricca Chemical

Chemical Identifiers

Chemical Name or Material Methanol
Name Note for Anethole Blank
CAS 67-56-1
CAS Min % 98.0
CAS Max % 100.0
Purity Grade HPLC Grade
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

R48280001C

 
ricca chemical company
RIR48280001C
Glass amber 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R48280004C

 
ricca chemical company
RIR48280004C
Glass amber 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Purity Grade HPLC Grade
Color Clear colorless
Quantity 1L
CAS 67-56-1
CAS Max % 100.0
CAS Min % 98.0
Packaging Glass amber
Chemical Name or Material Methanol
Name Note for Anethole Blank