Please login to your online account to display your discounted pricing

Methanol, HPLC Grade, for Anethole Blank, Ricca Chemical


Quantity 1L
Packaging Glass amber

Manufacturer: ricca chemical company  RIR48280001C

 View more versions of this product

Catalog No. R48280001C

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Purity Grade HPLC Grade
Physical Form Liquid
Color Clear colorless
Quantity 1L
CAS 67-56-1
Chemical Name or Material Methanol
Packaging Glass amber
Name Note for Anethole Blank
CAS Min % 98.0
CAS Max % 100.0

Specifications

Chemical Name or Material Methanol
Name Note for Anethole Blank
CAS 67-56-1
CAS Min % 98.0
CAS Max % 100.0
Purity Grade HPLC Grade
Physical Form Liquid