Please login to your online account to display your discounted pricing

Methanol For spectroscopy Uvasol™, EMD Millipore

For spectroscopy Uvasol™

$68.51 - $245.63

Chemical Identifiers

Chemical Name or Material Methanol
CAS 67-56-1
Molecular Formula CH4O
Formula Weight 32.04
UN Number 1230
Synonym Methyl alcohol, Carbinol, MeOH
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

M1060020500

 
emd millipore corporation
1060020500
Glass Bottle 500mL Each for $68.51

M1060022500

 
emd millipore corporation
1060022500
Glass Bottle 2.5L Each for $245.63
Description & Specifications

Specifications

Boiling Point 64.5°C
Melting Point −98°C
Formula Weight 32.04
Quantity 500mL
Chemical Name or Material Methanol
Ignition Point 455°C (851°F)
Odor Charactristic
Packaging Glass Bottle
Storage Information 5°C to 30°C (41°F to 86°F); Keep container tightly closed in a dry and well-ventilated place. Keep away from heat and sources of ignition. Keep locked up or in an area accessible only to qualified or authorized persons.
Vapor Pressure 128 hPa (20°C)
Flash Point 10°C (50°F)
Physical Form Liquid
Color Colorless
Molecular Formula CH4O
CAS 67-56-1
UN Number 1230
Synonym Methyl alcohol, Carbinol, MeOH
Solubility Information Soluble in water at 20°C
Health Hazard 1 H225: Highly flammable liquid and vapour.
Health Hazard 2 H301 + H311 + H331: Toxic if swallowed, in contact with skin or if inhaled
Description The Uvasol™ solvents range has been specially designed for spectroscopy and other applications requiring solvents of the highest spectral purity. The Uvasol™ solvents offer best UV transmittance.
Density 0.79g/cm3 at 20°C
Health Hazard 3 H370: Causes damage to organs
Safety and Handling

H225: Highly flammable liquid and vapour.

H370: Causes damage to organs

H301 + H311 + H331: Toxic if swallowed, in contact with skin or if inhaled