missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methanol For spectroscopy Uvasol™, MilliporeSigma™
For spectroscopy Uvasol™
Supplier: Technidata 1060020500
Specifications
Methanol | |
67-56-1 | |
0.79g/cm3 at 20°C | |
500 mL | |
455°C (851°F) | |
Glass Bottle | |
CH4O | |
128 hPa (20°C) | |
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
methanol | |
887 | |
32.04 |
-98°C | |
Colorless | |
64.5°C | |
10°C (50°F) | |
Charactristic | |
Liquid | |
MFCD00004595 | |
1230 | |
Soluble in water at 20°C | |
CO | |
32.04 | |
CHEBI:17790 |
Chemical Identifiers
67-56-1 | |
32.04 | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
887 | |
methanol |
CH4O | |
MFCD00004595 | |
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
CHEBI:17790 | |
CO |
Safety and Handling
H301 + H311 + H331: Toxic if swallowed, in contact with skin or if inhaled
H370: Causes damage to organs
H225: Highly flammable liquid and vapour.
Recommended Storage : 5°C to 30°C (41°F to 86°F); Keep container tightly closed in a dry and well-ventilated place. Keep away from heat and sources of ignition. Keep locked up or in an area accessible only to qualified or authorized persons.