Please login to your online account to display your discounted pricing

Methanol, For HPLC analysis, J.T.Baker™

High purity solvents for use in analytical and preparative separations

$136.25 - $302.08

Chemical Identifiers

Chemical Name or Material Methanol
CAS 67-56-1
Name Note For HPLC analysis
Purity Grade HPLC Grade
Molecular Formula CH4O
Formula Weight 32.04
Synonym Hydroxymethane; Methyl hydrate
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003342

 
avantor performance
909302
Narrow Mouth Amber Glass Bottle 1L Each for $136.25

02003349

 
avantor performance
909333
Narrow Mouth PVC Coated Amber Glass Bottle 4L Each for $302.08
Description & Specifications

Specifications

Formula Weight 32.04
Chemical Name or Material Methanol
Name Note For HPLC analysis
Purity Grade HPLC Grade
Melting Point −97.6°C
Molecular Formula CH4O
Synonym Hydroxymethane; Methyl hydrate
Density 0.7918g/cm3
Packaging Narrow Mouth Amber Glass Bottle
Flash Point 12°C
Boiling Point 64.7°C
Physical Form Liquid
Color Clear Colorless
Quantity 1L
CAS 67-56-1
Specific Gravity 1L = 0.80kg