Please login to your online account to display your discounted pricing

Methanol, Filtered through a 0.2 micron filter, BAKER ANALYZED™ LC-MS Reagent, J.T.Baker™

Function tested and optimized for LC/MS applications

$134.25 - $263.78

Chemical Identifiers

Chemical Name or Material Methanol
CAS 67-56-1
Name Note Filtered through a 0.2 micron filter
Purity Grade BAKER ANALYZED™ LC-MS Reagent
Molecular Formula CH3OH
Formula Weight 32.04
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003341

 
avantor performance
983003
Narrow Mouth Amber Glass Bottle 4L Each for $263.78

02003338

 
avantor performance
983002
Narrow Mouth Amber Glass Bottle 1L Each for $134.25
Description & Specifications

Specifications

Chemical Name or Material Methanol
Name Note Filtered through a 0.2 micron filter
CAS 67-56-1
Purity Grade BAKER ANALYZED™ LC-MS Reagent
Molecular Formula CH3OH
Formula Weight 32.04
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.80kg
Flash Point 12.00°C
Quantity 4L

    For Use in Liquid Chromatography and Mass Spectrometry; For Laboratory, Research, or Manufacturing Use