Please login to your online account to display your discounted pricing

Methanol-d4, for NMR, contains 0.03 v/v% TMS, 99.8% atom D, ACROS Organics™


Quantity 50mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  329920500

 View more versions of this product

Catalog No. AC329920500

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Methanol-d4
Name Note 99.8 atom % D
CAS 811-98-3
Deuterium (D) 99.75% min.
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Causes damage to organs.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Highly flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Immediately call a POISON CENTER or doctor/physician.
IF exposed: Call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
EINECS Number 212-378-6
TSCA TSCA
Water 0.05% max. (Coulometric)
Density 0.8880g/mL
Purity Grade For NMR
Physical Form Liquid
Quantity 50mL
Color Clear colorless
Molecular Formula CD4O
Linear Formula CD3OD
Formula Weight 36.06
MDL Number MFCD00044637
Beilstein 01, III, 1187

Specifications

Chemical Name or Material Methanol-d4
Name Note 99.8 atom % D
CAS 811-98-3
Purity Grade For NMR
Molecular Formula CD4O
Linear Formula CD3OD
Formula Weight 36.06
MDL Number MFCD00044637
Accessories

Danger

GHS P Statement:
Immediately call a POISON CENTER or doctor/physician.
IF exposed: Call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes damage to organs.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Highly flammable liquid and vapor.

SDS & Safety Documents