Please login to your online account to display your discounted pricing

Methanol-d4, for NMR, contains 0.03 v/v% TMS, 99.8% atom D, ACROS Organics™

$121.13 - $197.53

Chemical Identifiers

Chemical Name or Material Methanol-d4
Name Note 99.8 atom % D
CAS 811-98-3
Purity Grade For NMR
Molecular Formula CD4O
Linear Formula CD3OD
Formula Weight 36.06
MDL Number MFCD00044637
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC329920050

  SDS
thermo fisher scientific
329920050
Glass bottle 5mL Each for $121.13

AC329920500

  SDS
thermo fisher scientific
329920500
Glass bottle 50mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC329920100

  SDS
thermo fisher scientific
329920100
Glass bottle 10mL Each for $197.53
Description & Specifications

Specifications

Formula Weight 36.06
Physical Form Liquid
Quantity 5mL
Color Clear colorless
TSCA TSCA
EINECS Number 212-378-6
Purity Grade For NMR
Density 0.8880g/mL
Beilstein 01, III, 1187
Packaging Glass bottle
Water 0.05% max. (Coulometric)
Health Hazard 3 GHS P Statement:
Immediately call a POISON CENTER or doctor/physician.
IF exposed: Call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00044637
Deuterium (D) 99.75% min.
Health Hazard 2 GHS H Statement:
Causes damage to organs.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Highly flammable liquid and vapor.
Health Hazard 1 Danger
Chemical Name or Material Methanol-d4
Molecular Formula CD4O
Name Note 99.8 atom % D
Linear Formula CD3OD
CAS 811-98-3

For NMR

Safety and Handling

Danger

GHS P Statement:
Immediately call a POISON CENTER or doctor/physician.
IF exposed: Call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes damage to organs.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Highly flammable liquid and vapor.