Please login to your online account to display your discounted pricing

Methanol - D4, deuteration degree min. 99.95% for NMR spectroscopy MagniSolv™, EMD Millipore™


For NMR Spectroscopy

Quantity 10 x 0.5mL
Packaging Glass ampoule

Manufacturer: emd millipore corporation  1060250005

 View more versions of this product

Catalog No. M1060250005

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 36.07
Quantity 10 x 0.5mL
Color Colorless
Viscosity 0.597mPa&middnot;s (20°C/68°F)
Physical Form Liquid
Assay Percent Range 99.95% min.
Chemical Name or Material Methanol - D4
Vapor Pressure 128 hPa at 20°C (68°F)
Ignition Point 455°C (851°F)
Molecular Formula CD3OD
CAS 811-98-3
Synonym Tetradeuteromethanol
Odor Characteristic
Packaging Glass ampoule

Specifications

Chemical Name or Material Methanol - D4
CAS 811-98-3
Physical Form Liquid
Assay Percent Range 99.95% min.
Molecular Formula CD3OD
Formula Weight 36.07
Synonym Tetradeuteromethanol